missing translation for 'onlineSavingsMsg'
Learn More

TNFAIP1 Antibody - BSA Free, Novus Biologicals™

Product Code. 18398190 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398190 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18398190 Supplier Novus Biologicals Supplier No. NBP282039

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TNFAIP1 Polyclonal specifically detects TNFAIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFAIP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias B12, B61, BACURD2, BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 2, BTB/POZ domain-containing protein TNFAIP1, EDP1, hBACURD2, MGC2317, Protein B12, tumor necrosis factor, alpha-induced protein 1 (endothelial), Tumor necrosis factor, alpha-induced protein 1, endothelial
Gene Symbols TNFAIP1
Host Species Rabbit
Immunogen TNFAIP1 antibody was raised against a 19 amino acid peptide from near the carboxy terminus of human TNFAIP1. The immunogen is located within the last 50 amino acids of TNFAIP1. Amino Acid Squence: KRYSTYDDRQLGHQSTHRD
Molecular Weight of Antigen 35 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 7126
Test Specificity TNFAIP1 antibody is human, mouse and rat reactive. TNFAIP1 antibody is predicted to not cross-react with other TNFAIP proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.