missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF RII/TNFRSF1B Antibody (RM0012-6F13), DyLight 594, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NB110-85470DL594
This item is not returnable.
View return policy
Description
TNF RII/TNFRSF1B Monoclonal antibody specifically detects TNF RII/TNFRSF1B in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF
Specifications
| TNF RII/TNFRSF1B | |
| Monoclonal | |
| DyLight 594 | |
| CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNF-RII, TNF-R-II, TNFR-II, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II | |
| Mouse TNF RII/TNFRSF1B (extracellular domain) | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Signal Transduction, Zinc Finger | |
| 7133 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF | |
| RM0012-6F13 | |
| 50mM Sodium Borate | |
| Armenian Hamster | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction