missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TNF RII/TNFRSF1B Antibody Pair [HRP]

Product Code. p-7183321 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18274438

Brand: Novus Biologicals™ NBP27953015PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse TNF RII/TNFRSF1B.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 9.4 pg/mL
Assay Sensitivity 9.38-600 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse TNF RII/TNFRSF1B.
Target Species Mouse
For Use With (Application) Sandwich ELISA
Gene Alias CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNF-RII, TNF-R-II, TNFR-II, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II
Gene ID (Entrez) 7133
Gene Symbol TNFRSF1B
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 7 ng of recombinant mouse TNF RII/TNFRSF1B.
  • Capture Antibody: 1.0 mg/mL of rabbit anti-mouse TNF RII/TNFRSF1B monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.5 mg/mL rabbit anti-TNF RII/TNFRSF1B monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Signal Transduction, Zinc Finger
    Storage Requirements Storage is content dependent.
    Target TNF RII/TNFRSF1B
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.6 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.