missing translation for 'onlineSavingsMsg'
Learn More

TNF RII/TNFRSF1B Antibody (340), DyLight 488, Novus Biologicals™

Product Code. 18327610 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327610 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327610 Supplier Novus Biologicals Supplier No. NBP290436G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TNF RII/TNFRSF1B Monoclonal specifically detects TNF RII/TNFRSF1B in Mouse samples. It is validated for Sandwich ELISA Detection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF RII/TNFRSF1B
Applications ELISA
Classification Monoclonal
Clone 340
Conjugate DyLight 488
Dilution Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNF-RII, TNF-R-II, TNFR-II, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TNF RII/TNFRSF1B (NP_035740.2; Met1-Gly258).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Signal Transduction, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7133
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.