missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF RI/TNFRSF1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£240.00 - £353.00
Specifications
| Antigen | TNF RI/TNFRSF1A |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20-1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18435731
|
Novus Biologicals
NBP1-88140-25ul |
25 μL |
£240.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18265446
|
Novus Biologicals
NBP1-88140 |
0.1 mL |
£353.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TNF RI/TNFRSF1A Polyclonal specifically detects TNF RI/TNFRSF1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TNF RI/TNFRSF1A | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNF-RI, TNF-R-I, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor | |
| TNFRSF1A | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20-1:50 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Cytokine Research | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 7132 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GDREKRDSVCPQGKYIHPQNNSICCTKCHKGTYLYNDCPGPGQDTDCRECESGSFTASENHLRHCLSCSKCRKEMGQVEISSCTVDRDTVCGCRKNQYRHYWSENLFQCFNCSLCLNGTVHLSCQEKQNTVCTCHAGFFLRENECVSC | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title