missing translation for 'onlineSavingsMsg'
Learn More

PeproTech TNF beta Polyclonal Antibody, PeproTech®, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  PeproTech 500-P31B-50UG

 View more versions of this product

Product Code. 17835653

  • £177.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: MLPGVGLTPS AAQTARQHPK MHLAHSTLKP AAHLIGDPSK QNSLLWRANT DRAFLQDGFS LSNNSLLVPT SGIYFVYSQV VFSGKAYSPK ATSSPLYLAH EVQLFSSQYP FHVPLLSSQK MVYPGLQEPW LHSMYHGAAF QLTQGDQLST HTDGIPHLVL SPSTVFFGAF AL. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human TNF-beta. Anti-Human TNF-beta-specific antibody was purified by affinity chromatography employing an immobilized Human TNF-beta matrix. Sandwich ELISA: To detect Human TNF-beta by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human TNF-beta (500-P31BBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human TNF-beta. Western Blot: To detect Human TNF-beta by Western Blot analysis this antibody can be used at a concentration of 0.1- 0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human TNF-beta is 1.5-3.0 ng/lane, under either reducing non-reducing conditions.

TNF beta, a member of the tumor necrosis factor family, is a cytokine produced by lymphocytes. The protein is highly inducible, secreted, and forms heterotrimers with lymphotoxin-beta which anchor lymphotoxin-alpha to the cell surface. This protein also mediates a large variety of inflammatory, immunostimulatory, and antiviral responses, is involved in the formation of secondary lymphoid organs during development and plays a role in apoptosis. Genetic variations in this gene are associated with susceptibility to leprosy type 4 and psoriatic arthritis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TNF beta
Polyclonal
Unconjugated
LTA
DAMA-25N12.13; hlb382; hypothetical protein; LT; LT[a]; LT-[a]; LTA; LTalpha; LT-alpha; Ltx; lymphotoxin A; lymphotoxin alpha; lymphotoxin alpha (TNF superfamily, member 1); lymphotoxin-alpha; lymphotoxin-alpha; lymphotoxin alpha; TNF b; TNF beta; TNF superfamily, member 1; TNF β; Tnfb; TNFbeta; TNF-beta; TNF-N; Tnfsf1; Tnfsf1b; TNFβ; TNLG1E; tumor necrosis factor beta; tumor necrosis factor ligand 1E; tumor necrosis factor ligand superfamily member 1
Rabbit
Antigen affinity chromatography
RUO
4049
-20°C
Lyophilized
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
P01374
LTA
E.coli-derived Recombinant Human TNF-beta.
50 μg
Primary
Human
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.