Learn More
PeproTech TNF beta Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P31B-1MG
Description
AA Sequence of recombinant protein: MLPGVGLTPS AAQTARQHPK MHLAHSTLKP AAHLIGDPSK QNSLLWRANT DRAFLQDGFS LSNNSLLVPT SGIYFVYSQV VFSGKAYSPK ATSSPLYLAH EVQLFSSQYP FHVPLLSSQK MVYPGLQEPW LHSMYHGAAF QLTQGDQLST HTDGIPHLVL SPSTVFFGAF AL. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human TNF-beta. Anti-Human TNF-beta-specific antibody was purified by affinity chromatography employing an immobilized Human TNF-beta matrix. Sandwich ELISA: To detect Human TNF-beta by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human TNF-beta (500-P31BBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human TNF-beta. Western Blot: To detect Human TNF-beta by Western Blot analysis this antibody can be used at a concentration of 0.1- 0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human TNF-beta is 1.5-3.0 ng/lane, under either reducing non-reducing conditions.
TNF beta, a member of the tumor necrosis factor family, is a cytokine produced by lymphocytes. The protein is highly inducible, secreted, and forms heterotrimers with lymphotoxin-beta which anchor lymphotoxin-alpha to the cell surface. This protein also mediates a large variety of inflammatory, immunostimulatory, and antiviral responses, is involved in the formation of secondary lymphoid organs during development and plays a role in apoptosis. Genetic variations in this gene are associated with susceptibility to leprosy type 4 and psoriatic arthritis.
Specifications
| TNF beta | |
| Polyclonal | |
| Unconjugated | |
| LTA | |
| DAMA-25N12.13; hlb382; hypothetical protein; LT; LT[a]; LT-[a]; LTA; LTalpha; LT-alpha; Ltx; lymphotoxin A; lymphotoxin alpha; lymphotoxin alpha (TNF superfamily, member 1); lymphotoxin-alpha; lymphotoxin-alpha; lymphotoxin alpha; TNF b; TNF beta; TNF superfamily, member 1; TNF β; Tnfb; TNFbeta; TNF-beta; TNF-N; Tnfsf1; Tnfsf1b; TNFβ; TNLG1E; tumor necrosis factor beta; tumor necrosis factor ligand 1E; tumor necrosis factor ligand superfamily member 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4049 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P01374 | |
| LTA | |
| E.coli-derived Recombinant Human TNF-beta. | |
| 1 mg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.