missing translation for 'onlineSavingsMsg'
Learn More

PeproTech TNF alpha Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  PeproTech 500-P72BT-50UG

 View more versions of this product

Product Code. 17863593

  • £228.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: MLRSSSQNSS DKPVAHVVAN HQAEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLIY SQVLFKGQGC PDYVLLTHTV SRFAISYQEK VSLLSAIKSP CPKDTPEGAE LKPWYEPMYL GGVFQLEKGD LLSAEVNLPK YLDITESGQV YFGVIAL. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat TNF-alpha. Anti-Rat TNF-alpha-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Rat TNF-alpha by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Rat TNF-alpha (500-P72) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat TNF-alpha. Western Blot: To detect Rat TNF-alpha by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Rat TNF-alpha is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P72BT-1MG will be provided as 2 x 500 μg

TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TNF alpha
Polyclonal
Biotin
Tnf
APC1 protein; Cachectin; C-domain 1; C-domain 2; cTNF; DADB-70P7.1; DIF; ICD1; ICD2; Intracellular domain 1; Intracellular domain 2; N-terminal fragment; NTF; RATTNF; Tnf; TNF alpha; TNF superfamily; TNF α; TNF, macrophage-derived; TNF, monocyte-derived; TNFA; TNF-a; TNFalpha; TNF-alpha; Tnfsf1a; TNFSF2; TNFα; TNLG1F; Tumor necrosis factor; tumor necrosis factor (TNF superfamily, member 2); tumor necrosis factor alpha; tumor necrosis factor alpha (cachetin); tumor necrosis factor alpha precursor; tumor necrosis factor ligand 1F; tumor necrosis factor ligand superfamily member 2; Tumor necrosis factor, membrane form; Tumor necrosis factor, soluble form; tumor necrosis factor-alpha; tumor necrosis factor-alpha precursor; tumor-necrosis factor; tumour necrosis factor
Rabbit
Antigen affinity chromatography
RUO
24835
-20°C
Lyophilized
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
P16599
Tnf
E.coli-derived, 17.3 kDa Recombinant Rat TNF-alpha.
50 μg
Primary
Rat
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.