missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TNF alpha Mouse Uncoated ELISA Kit

Uncoated ELISA Kit

£367.00 - £491.00

Specifications

Accession Number P06804
Assay Range 8-1,000 pg/mL
Assay Sensitivity 8 pg/mL
Conjugate Biotin
Product Type ELISA Kit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15581107
View Documents Promotion Details
Invitrogen™
88-7324-77
20 x 96 Tests
£491.00
Pack of 20
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
15501117
View Documents Promotion Details
Invitrogen™
88-7324-88
10 x 96 Tests
£367.00
Pack of 10
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
Description

Description

The Mouse Tumor Necrosis Factor Alpha (TNF-α) Uncoated ELISA Kit contains pre-matched antibody pairs, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse TNF-α. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Tumor Necrosis Factor is a polypeptide cytokine produced by monocytes and macrophages. It functions as a multipotent modulator of immune response and further acts as a potent pyrogen. TNF-alpha circulates throughout the body responding to stimuli (infectious agents or tissue injury), activating neutrophils, altering the properties of vascular endothelial cells, regulating metabolic activities of other tissues, as well as exhibiting tumoricidal activity by inducing localized blood clotting. TNF-alpha also inhibits lipoprotein lipase activity resulting in cachexia, a physical wasting condition. Activation of B-cells by the Epstein Barr virus can be inhibited by TNF-alpha. Due to its varied actions throughout the immune system, TNF-alpha may play a role in the pathogenesis of many disease states. TNF-alpha production is mediated by the action of lymphokines and endotoxins on the macrophage. Purified monocytes produce TNF-alpha within four hours of stimulation by recombinant IL-2 and there is some in vitro evidence to suggest that TNF-alpha is expressed at high levels and with prolonged kinetics in T cells stimulated by both CD2 and CD28. Secretion of TNF-alpha is enhanced by gamma interferon. TNF then induces or enhances the specific production of Class I MHC antigen, GM-CSF, and IL-1.
Specifications

Specifications

P06804
8 pg/mL
ELISA Kit
Mouse
Colorimetric Microplate Reader
AA690181,IL-15RA,AI503618,IL-15
HRP
Lavage Fluid,100 μL; Plasma, 100 μL; Serum, 100 μL; Supernatant, 100 μL; Tissue Homogenate,100 μL
Tumor Necrosis Factor alpha, tnf-a,tnf alpha
1 hr. 45 min.
8-1,000 pg/mL
Biotin
Lavage Fluid, Plasma, Serum, Supernatant, Tissue Homogenates
ELISA
21926
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant cytokine for generating standard curve and calibrating samples, ELISA/ELISPOT Coating Buffer Powder: This uncoated ELISA kit may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 μM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20)., Assay Diluent: 5X concentrated, Detection enzyme: Pre-titrated Avidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -44; -76; -86)
RUO
2°C to 8°C
Mouse
24 hr. 30 min.
Videos
SDS
Documents

Documents

Product Certifications