missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF-alpha Mouse anti-Human, Clone: B-F7, Novus Biologicals™
Mouse Monoclonal Antibody
£319.00 - £603.00
Specifications
| Antigen | TNF-alpha |
|---|---|
| Clone | B-F7 |
| Concentration | 1 mg/mL |
| Dilution | ELISA, ELISPOT |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18352586
|
Novus Biologicals
NBP3-14590-200UL |
200 μg |
£319.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18362542
|
Bio-Techne
NBP3-14590-500UL |
500 μg |
£603.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TNF-alpha Monoclonal antibody specifically detects TNF-alpha in Human samples. It is validated for ELISA, ELISPOTSpecifications
| TNF-alpha | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| PBS. Sterile-filtered through 0.22 um. | |
| 7124 | |
| IgG1 κ | |
| Affinity purified | |
| Antibody |
| B-F7 | |
| ELISA, ELISPOT | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Recombinant human TNF-alpha | |
| Primary | |
| Store at 4°C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title