missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF-alpha Antibody (D2E7 (Adalimumab)), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
TNF-alpha Monoclonal antibody specifically detects TNF-alpha in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Neutralization
Specifications
Specifications
| Antigen | TNF-alpha |
| Applications | Neutralization |
| Purification Method | Protein A purified |
| Regulatory Status | RUO |
| Research Discipline | Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity |
| Primary or Secondary | Primary |
| Isotype | IgG κ |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?