Learn More
TNF-alpha Antibody (D2E7 (Adalimumab)) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP2-62567
 View more versions of this product
Product Code. 18619506
  • £494.00 / 0.20mg
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Block/Neutralize.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TNF-alpha
Monoclonal
Unconjugated
PBS
TNF
This antibody was generated by affinity maturation from the 2SD4 scFv, which in turn was selected on human TNF alpha by guided phage-display technology, using the murine anti-hTNF alpha antibody Mab32 as a template (c.f. also US Patent 6090382).
7124
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
IgG1 κ
Neutralization, ELISA, Immunohistochemistry, Blocking Assay, Immunofluorescence
D2E7 (Adalimumab)
ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunofluorescence 1:10 - 1:500, Block/Neutralize
APC1 protein, biosimilar, biosimilars, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Human
0.2 mg
Binds to soluble TNF-alpha, but not to lymphotoxin (TNF beta).
Purified