missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF-alpha Antibody (CDP 571 (Humicade)) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-52679-0.025MG
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Neutralization.
Specifications
| TNF-alpha | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| TNF | |
| Human TNF-alpha | |
| 0.025 mg | |
| Adaptive Immunity, Asthma, Autophagy, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| 7124 | |
| Human | |
| Purified |
| ELISA | |
| CDP 571 (Humicade) | |
| ELISA 1:100 - 1:2000 | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| The antibody binds to human TNF-alpha with a Kd of 0.087 nM. | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction