missing translation for 'onlineSavingsMsg'
Learn More

TNF-alpha Antibody (CDP 571 (Humicade)), Biotin, Novus Biologicals™

Product Code. 18386770 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386770 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386770 Supplier Novus Biologicals Supplier No. NBP275908B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF-alpha
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone CDP 571 (Humicade)
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Gene Symbols TNF
Host Species Human
Immunogen Human TNF-alpha
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 7124.0
Test Specificity The antibody binds to human TNF-alpha with a Kd of 0.087 nM.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG4 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.