missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF-alpha Antibody (cA2 (Infliximab)), Allophycocyanin - Chimeric, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP2-75898APC
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Flow Cytometry, Flow (Intracellular).
Specifications
| TNF-alpha | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| TNF | |
| Purified recombinant human TNF (rTNF). | |
| 0.1 mL | |
| Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| Binds to soluble and transmembrane forms of TNF-alpha and neutralizes its biological activity. Does not bind to TNF-beta. | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| cA2 (Infliximab) | |
| Flow Cytometry, Flow (Intracellular) | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction