missing translation for 'onlineSavingsMsg'
Learn More

TNF-alpha Antibody (6N2E12) - Azide and BSA Free, Novus Biologicals™

Product Code. 18389210 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389210 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389210 Supplier Novus Biologicals Supplier No. NBP280984

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Western Blot, Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF-alpha
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 6N2E12
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4 μg/mL, Flow Cytometry : 0.5-2ug/10^6cells
Formulation PBS
Gene Alias APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Gene Symbols TNF
Host Species Mouse
Immunogen Full-length recombinant human TNF alpha was used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 7124
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.