missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF-alpha Antibody (2342C) - Low Endotoxin, Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-75925
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Neutralization.
Specifications
| TNF-alpha | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 2 μg/mL, Immunohistochemistry-Paraffin, Neutralization | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alpha, TNF-alpha/TNFSF2, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| 7124.0 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin), Neutralization | |
| 2342C | |
| Unconjugated | |
| PBS with No Preservative | |
| TNF | |
| Partial recombinant human TNF-alpha protein (amino acids 77-233) [Uniprot: P01375] | |
| 0.1 mg | |
| Primary | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction