missing translation for 'onlineSavingsMsg'
Learn More

TN1, MedChemExpress

Product Code. 16493178
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493178 10 mg 10mg
16434466 100 mg 100mg
16415917 5 mg 5mg
16410709 1 mg 1mg
16409736 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493178 Supplier Medchem Express Supplier No. HY100826
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress TN1 is a potent fetal hemoglobin (HbF) inducer.

TN1 is a potent fetal hemoglobin (HbF) inducer.

Specifications

Chemical Name or Material TN1
CAS 289479-94-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H31N7O2
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (196.23 mM; Need ultrasonic)
SMILES O=C(NC1=CC=CC(NC2=C3N=CN(CC)C3=NC(N[C@H]4CC[C@H](O)CC4)=N2)=C1)C#CC5=CC=C(C)C=C5
Molecular Weight (g/mol) 509.6
Formula Weight 509.6
Percent Purity 95.14%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.