missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMX4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21348-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to TMX4.
This gene encodes a member of the disulfide isomerase (PDI) family of endoplasmic reticulum (ER) proteins that catalyze protein folding and thiol-disulfide interchange reactions. The encoded protein has an N-terminal ER-signal sequence, a catalytically active thioredoxin domain, one transmembrane domain and C-terminal ASP/GLU-rich calcium binding domain. Unlike most members of this gene family, it lacks a C-terminal ER-retention sequence. The encoded protein has been shown to have reductase activity in vitro.Specifications
| TMX4 | |
| Polyclonal | |
| Unconjugated | |
| TMX4 | |
| KIAA1162, PDIA14, TMX4, TXNDC13, UNQ475/PRO938 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 296182, 52837, 56255 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.56 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8C0L0, Q9H1E5 | |
| Tmx4 | |
| TMX4 Fusion Protein Ag15696 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction