missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMX4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551380
This item is not returnable.
View return policy
Description
Immunogen sequence: QLTALLAAWI AAVAATAGPE EAALPPEQSR VQPMTASNWT LVMEGEWMLK FYAPWCPSCQ QTDSEWEAFA KNGEILQISV GKVDVIQEPG LSGRFFVTTL PAFFHAKDGI F Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
This gene encodes a member of the disulfide isomerase (PDI) family of endoplasmic reticulum (ER) proteins that catalyze protein folding and thiol-disulfide interchange reactions. The encoded protein has an N-terminal ER-signal sequence, a catalytically active thioredoxin domain, one transmembrane domain and C-terminal ASP/GLU-rich calcium binding domain. Unlike most members of this gene family, it lacks a C-terminal ER-retention sequence. The encoded protein has been shown to have reductase activity in vitro.
Specifications
| TMX4 | |
| Polyclonal | |
| Unconjugated | |
| TMX4 | |
| 2810417D04Rik; 4930500L08Rik; AI843224; AW046784; D2Bwg1356e; DJ971N18.2; Kiaa1162; mKIAA1162; PDIA14; protein disulfide isomerase family A, member 14; PSEC0095; RGD1305146; thioredoxin domain containing 13; thioredoxin domain-containing protein 13; thioredoxin related transmembrane protein 4; thioredoxin-related transmembrane protein 4; Tmx4; Txndc13; UNQ475/PRO938 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56255 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H1E5 | |
| TMX4 | |
| Recombinant protein corresponding to Human TMX4. Recombinant protein control fragment (Product #RP-93684). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction