missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19838-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the disulfide isomerase (PDI) family of endoplasmic reticulum (ER) proteins that catalyze protein folding and thiol-disulfide interchange reactions. The encoded protein has an N-terminal ER-signal sequence, a catalytically active thioredoxin domain, one transmembrane domain and a C-terminal ER-retention sequence. This protein is enriched on the mitochondria-associated-membrane of the ER via palmitoylation of two of its cytosolically exposed cysteines.Specifications
| TMX2 | |
| Polyclonal | |
| Unconjugated | |
| TMX2 | |
| CGI 31, My009, PDIA12, PIG26, PSEC0045, TMX2, TXNDC14, UNQ237/PRO270 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 295701, 51075, 66958 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5XIK2, Q9D710, Q9Y320 | |
| TMX2 | |
| TMX2 Fusion Protein Ag13828 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction