missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109348
This item is not returnable.
View return policy
Description
Positive Control: Daudi cell lysates, MG-63, human skin tissue, human breast carcinoma tissue, F9.
TXNDC1 is a thioredoxin (TXN; see MIM 187700)-related protein with disulfide reductase activity (Matsuo et al., 2001 [PubMed 11152479]).
Specifications
| TMX | |
| Polyclonal | |
| Unconjugated | |
| TMX1 | |
| 2810425A04Rik; AI465319; DKFZP564E1962; PDIA11; protein disulfide isomerase family A, member 11; PSEC0085; thioredoxin domain containing 1; thioredoxin domain-containing protein 1; thioredoxin related transmembrane protein 1; thioredoxin-related transmembrane protein 1; TMX; Tmx1; Transmembrane Trx-related protein; TXNDC; TXNDC1; UNQ235/PRO268 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 81542 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9H3N1 | |
| TMX1 | |
| Synthetic peptide within Human TMX1 aa 89-138. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction