missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMX Monoclonal Antibody (OTI3H5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526309
This item is not returnable.
View return policy
Description
TMX Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
TXNDC1 is a thioredoxin (TXN; see MIM 187700)-related protein with disulfide reductase activity (Matsuo et al., 2001 [PubMed 11152479]).
Specifications
| TMX | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H3N1 | |
| TMX1 | |
| Full length human recombinant protein of TMX1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI3H5 | |
| Unconjugated | |
| TMX1 | |
| 2810425A04Rik; AI465319; DKFZP564E1962; PDIA11; protein disulfide isomerase family A, member 11; PSEC0085; thioredoxin domain containing 1; thioredoxin domain-containing protein 1; thioredoxin related transmembrane protein 1; thioredoxin-related transmembrane protein 1; TMX; Tmx1; Transmembrane Trx-related protein; TXNDC; TXNDC1; UNQ235/PRO268 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 81542 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction