missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TMTC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29438
This item is not returnable.
View return policy
Description
TMTC3 Polyclonal antibody specifically detects TMTC3 in Human samples. It is validated for Immunoprecipitation
Specifications
| TMTC3 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| DKFZp686C0968, DKFZp686M1969, DKFZp686O22167, DKFZp686O2342, FLJ90492, SMILE, transmembrane and tetratricopeptide repeat containing 3, transmembrane and TPR repeat-containing protein 3 | |
| between 775 and 825 | |
| 100 μg | |
| Cell Biology | |
| 160418 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction