missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMTC2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-09267-100UL
This item is not returnable.
View return policy
Description
TMTC2 Polyclonal specifically detects TMTC2 in Human samples. It is validated for Western Blot.
Specifications
| TMTC2 | |
| Polyclonal | |
| Western Blot 1.0 ug/ml | |
| DKFZp762A217, transmembrane and tetratricopeptide repeat containing 2, transmembrane and TPR repeat-containing protein 2 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of human TMTC2 (NP_689801.1). Peptide sequence YLNTGIILMNQGRTEEARRTFLKCSEIPDENLKDPHAHKSSVTSCLYNLG | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 160335 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction