missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMTC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555466
This item is not returnable.
View return policy
Description
Immunogen sequence: MMGEAYMRLS KLPEAEHWYM ESLRSKTDHI PAHLTYGKLL ALTGRKSEAE KLFLKAIELD PTKGNCYMHY GQFLLEEARL IEAAEMAKKA AELDSTEFDV VFNAAHMLRQ ASLNEAAEKY YDLAARLRPN YPAALMNLGA IL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 94%.
Specifications
| TMTC2 | |
| Polyclonal | |
| Unconjugated | |
| TMTC2 | |
| 8430438D04Rik; D330034A10Rik; IBDBP1; IBDBP1 transcript 2; Protein O-mannosyl-transferase TMTC2; RGD1309848; TMTC2; transmembrane and tetratricopeptide repeat containing 2; transmembrane and TPR repeat-containing protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 160335, 278279, 299762 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q56A06, Q8N394 | |
| TMTC2 | |
| Recombinant protein corresponding to Human TMTC2. Recombinant protein control fragment (Product #RP-90774). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction