missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMTC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23349-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| TMTC1 | |
| Polyclonal | |
| Unconjugated | |
| TMTC1 | |
| ARG99, OLF, TMTC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 387314, 83857 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3UV71, Q8IUR5 | |
| Tmtc1 | |
| TMTC1 Fusion Protein Ag19944 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction