missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMT Monoclonal Antibody (25D5)
GREENER_CHOICE

Product Code. 11858203
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11858203 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11858203 Supplier Invitrogen™ Supplier No. 90075

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Product # 90075 binds to immobilized TMT surface. Product # 90075 has been succesfully used in immunoprecipitation and Western blot applications.

Tandem Mass Tag (TMT) reagents enable concurrent identification and multiplexed quantitation of proteins in different samples using tandem mass spectrometry (MS). Isobaric TMT reagents share identical strucures, including a peptide-reactive labeling group, a spacer arm and a MS/MS reporter. During the MS/MS analysis, each isobaric tag produces a unique reporter ion that makes quantitation possible. Amine-reactive NHS-ester isotopic duplex or six-plex isobaric TMT tags covalently react with amino termini and lysine residues of peptides and proteins. Cysteine-reactive Tandem Mass Tag (cysTMT) reagents react with reduced thiols of cysteine-containing peptides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMT
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 25D5
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 4% BSA and Kathon
Gene Alias Tandem Mass Tag
Host Species Mouse
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.