missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMSB4X Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550584
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total Thymosin beta 4 protein.
Thymosin-beta4 is a small, actin-sequestering protein belonging to the thymosin-beta family that is found at high concentrations within the spleen, thymus, and peritoneal macrophages, where it is most notably responsible for the organization of cytoskeletal structure. In mammalian tissues, this protein acts as a modulator for the polymerization/depolymerization of actin through the formation of a 1:1 complex with the monomer G (globular)-actin, and inhibits actin's polymerization to form F (filamentous) actin, which together with other proteins binds microfilaments to construct the cytoskeleton. Commonly found at significant quantities within the brain, lungs, liver, kidneys, testes, and heart, thymosin-beta4 has also been shown to be synthesized by cells unrelated to the reticuloendothelial system, such as myoblasts and fibroblasts, and expressed at irregular levels by several hemopoietic cell lines, malignant lymphoid cells and myeloma cells. In addition to regulating actin polymerization, research has also found thymosin-beta4 to stimulate the secretion of hypothalamic luteinizing hormone-releasing hormone and luteinizing hormone, inhibit the migration of peritoneal macrophages, induce phenotypic changes in T cell lines during early host defense mechanisms, and inhibit the progression of hematopoietic pluripotent stem cells into the S-phase.
Specifications
| TMSB4X | |
| Polyclonal | |
| Unconjugated | |
| TMSB4X | |
| Fx; Hematopoietic system regulatory peptide; prothymosin beta 4; prothymosin beta-4; Ptmb4; Seraspenide; T beta 4; t beta-4; Tb4; TB4X; TB4Xbeta 4; Thyb4; thymosin; thymosin beta 4; thymosin beta 4, X-linked; thymosin beta-4; thymosin, beta 4, X chromosome; Tmsb4; Tmsb4x | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19241, 7114, 81814 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P20065, P62328, P62329 | |
| TMSB4X | |
| Synthetic peptide corresponding to residues near the N terminal of human thymosin beta 4, X-linked. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction