missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMSB10 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149202
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This protein plays an important role in the organization of the cytoskeleton. It binds to and sequesters actin monomers (G actin) and therefore inhibits actin polymerization. Experiment indicate that the expression decrease dramatically after birth; It expresses high levels in human fetal brain.
Specifications
| TMSB10 | |
| Polyclonal | |
| Unconjugated | |
| TMSB10 | |
| MIG12; migration-inducing gene 12; migration-inducing protein 12; prothymosin beta 10; Ptmb10; Tb10; Thyb10; thymosin; thymosin beta 10; thymosin beta-10; thymosin, beta 10; TMSB10 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 19240, 50665, 9168 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P63312, P63313, Q6ZWY8 | |
| TMSB10 | |
| E. coli-derived recombinant human Thymosin beta10, Ala2-Ser44. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction