missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMSB10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116041
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Thymosin beta 10.
This protein plays an important role in the organization of the cytoskeleton. It binds to and sequesters actin monomers (G actin) and therefore inhibits actin polymerization. Experiment indicate that the expression decrease dramatically after birth; It expresses high levels in human fetal brain.
Specifications
| TMSB10 | |
| Polyclonal | |
| Unconjugated | |
| TMSB10 | |
| MIG12; migration-inducing gene 12; migration-inducing protein 12; prothymosin beta 10; Ptmb10; Tb10; Thyb10; thymosin; thymosin beta 10; thymosin beta-10; thymosin, beta 10; TMSB10 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19240, 50665, 9168 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P63312, P63313, Q6ZWY8 | |
| TMSB10 | |
| A synthesized peptide derived from human TMSB10(Accession P63313), corresponding to amino acid residues M1-E36. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction