missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMPRSS9 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5149046

Product Code. 30534670

  • £357.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

The protein encoded by this gene is a membrane-bound type II serine polyprotease that is cleaved to release three different proteases. Two of the proteases are active and can be inhibited by serine protease inhibitors, and one is thought to be catalytically inactive. This gene enhances the invasive capability of pancreatic cancer cells and may be involved in cancer progression.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TMPRSS9
Polyclonal
Unconjugated
TMPRSS9
polyserase 1; polyserase-1; Polyserase-I; polyserine protease 1; polyserine protease-1; Serase-1; serase-1b; Serase-2; Serase-3; TMPRSS9; Transmembrane protease serine 9; transmembrane protease, serine 9; transmembrane serine protease 9
Goat
Antigen affinity chromatography
RUO
360200
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
Q7Z410
TMPRSS9
E. coli-derived recombinant human TMPRSS9, Glu190-Gln490.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.