missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMPRSS7 Polyclonal Antibody

Product Code. 15644705
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15644705 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15644705 Supplier Invitrogen™ Supplier No. PA550776

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total TMPRSS7 protein.

Serine protease which preferentially hydrolyzes peptides with Arg at the P1 position. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMPRSS7
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TMPRSS7
Gene Accession No. Q7RTY8
Gene Alias B230219I23Rik; Gm1748; matriptase-3; TMPRSS7; Transmembrane protease serine 7; transmembrane protease, serine 7; transmembrane serine protease 7; type II transmembrane serine protease 7
Gene Symbols TMPRSS7
Host Species Rabbit
Immunogen Fusion protein corresponding to residues near the C terminal of human transmembrane protease, serine 7.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 344805
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.