missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMPRSS4 Rabbit anti-Human, Unconjugated, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5110705
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: DVFNWKVRAG SDKLGSFPSL AVAKIIIIEF NPMYPKDNDI ALMKLQFPLT FSGTVRPICL PFFDEELTPA TPLWIIGWGF TKQNGGKMSD ILLQASVQVI DSTRCNADDA YQGEVTEKMM
This gene encodes a member of the serine protease family. Serine proteases are known to be involved in a variety of biological processes, whose malfunction often leads to human diseases and disorders. This gene was identified as a gene overexpressed in pancreatic carcinoma. The encoded protein is membrane bound with a N-terminal anchor sequence and a glycosylated extracellular region containing the serine protease domain. Multiple transcript variants encoding different isoforms have been found for this gene.
Spezifikation
| TMPRSS4 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NRS4 | |
| TMPRSS4 | |
| Recombinant Protein corresonding to Human TMPRSS4 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| TMPRSS4 | |
| Cap2; CAPH2; Channel-activating protease 2; mCAP2; Membrane-type serine protease 2; MT-SP2; TMPRSS3; Tmprss4; Transmembrane protease serine 4; UNQ776/PRO1570 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56649 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur