missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMPRSS4 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11283-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the serine protease family. Serine proteases are known to be involved in a variety of biological processes, whose malfunction often leads to human diseases and disorders. This gene was identified as a gene overexpressed in pancreatic carcinoma. The encoded protein is membrane bound with a N-terminal anchor sequence and a glycosylated extracellular region containing the serine protease domain. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| TMPRSS4 | |
| Polyclonal | |
| Unconjugated | |
| TMPRSS4 | |
| CAPH2, Channel activating protease 2, MT SP2, TMPRSS3, TMPRSS4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 367074, 56649 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NRS4 | |
| Tmprss4 | |
| TMPRSS4 Fusion Protein Ag1804 | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction