missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMPRSS4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13494749
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13494749 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13494749 Supplier Invitrogen™ Supplier No. PA533084

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Chimpanzee, Orangutan, Gibbon, Monkey, Marmoset (100%) Mouse, Guinea pig (94%).

This gene encodes a member of the serine protease family. Serine proteases are known to be involved in a variety of biological processes, whose malfunction often leads to human diseases and disorders. This gene was identified as a gene overexpressed in pancreatic carcinoma. The encoded protein is membrane bound with a N-terminal anchor sequence and a glycosylated extracellular region containing the serine protease domain. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMPRSS4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene TMPRSS4
Gene Accession No. Q9NRS4
Gene Alias Cap2; CAPH2; Channel-activating protease 2; mCAP2; Membrane-type serine protease 2; MT-SP2; TMPRSS3; Tmprss4; Transmembrane protease serine 4; transmembrane protease, serine 4; transmembrane serine protease 3; type II membrane serine protease; UNQ776/PRO1570
Gene Symbols TMPRSS4
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from internal region of human TMPRSS4.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56649
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.