missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMPRSS3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575286
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a protein that belongs to the serine protease family. The encoded protein contains a serine protease domain, a transmembrane domain, a LDL receptor-like domain, and a scavenger receptor cysteine-rich domain. Serine proteases are known to be involved in a variety of biological processes, whose malfunction often leads to human diseases and disorders. This gene was identified by its association with both congenital and childhood onset autosomal recessive deafness. This gene is expressed in fetal cochlea and many other tissues, and is thought to be involved in the development and maintenance of the inner ear or the contents of the perilymph and endolymph. This gene was also identified as a tumor associated gene that is overexpressed in ovarian tumors. Alternatively spliced transcript variants have been described.
Specifications
| TMPRSS3 | |
| Polyclonal | |
| Unconjugated | |
| Tmprss3 | |
| DFNB10; DFNB8; ECHOS1; Serine protease TADG-12; TADG12; TMPRSS3; Transmembrane protease serine 3; transmembrane protease, serine 3; transmembrane proteinase serine 3; transmembrane serine protease 3; tumor-associated differentially-expressed gene 12 protein; UNQ323/PRO382 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140765, 309665, 64699 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P57727, Q8K1T0 | |
| Tmprss3 | |
| Synthetic peptide corresponding to amino acids 420-460 of Human TMPRSS3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction