missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TMPRSS2 Antibody (Regeneron patent anti-TMPRSS2) - Humanized, Novus Biologicals™
Shop All Bio Techne ProductsDescription
TMPRSS2 Monoclonal antibody specifically detects TMPRSS2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
Specifications
| Antigen | TMPRSS2 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Clone | Regeneron patent anti-TMPRSS2 |
| Conjugate | Unconjugated |
| Formulation | Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) |
| Gene Accession No. | O15393 |
| Gene Alias | EC 3.4.21, FLJ41954, NP_001128571.1, NP_001369649.1, NP_005647.3, PP9284, PRSS10, Serine protease 10, TMPRSS2, transmembrane protease serine 2, transmembrane protease, serine 2 |
| Host Species | Human |
| Immunogen | TMPRSS2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?