missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMPRSS2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-20984
This item is not returnable.
View return policy
Description
TMPRSS2 Polyclonal specifically detects TMPRSS2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| TMPRSS2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| EC 3.4.21, FLJ41954, NP_001128571.1, NP_001369649.1, NP_005647.3, PP9284, PRSS10, Serine protease 10, TMPRSS2, transmembrane protease serine 2, transmembrane protease, serine 2 | |
| Goat | |
| 53.9 kDa | |
| 0.1 mg | |
| Cancer, Cell Biology, Neuroscience, Prostate Cancer, Virology Bacteria and Parasites | |
| 7113 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.5 ug/mL, Simple Western, Peptide ELISA Detection limit 1:8000 | |
| NP_005647.2 | |
| TMPRSS2 | |
| This TMPRSS2 Antibody was developed against a peptide with sequence C-TDWIYRQMRADG corresponding to internal region according to NP_005647.2. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Dog. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction