missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMPRSS15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584912
This item is not returnable.
View return policy
Description
Immunogen sequence: LNDDNEWERI QGSTFSPFTG PNFDHTFGNA SGFYISTPTG PGGRQERVGL LSLPLDPTLE PACLSFWYHM YGENVHKLSI NISNDQNMEK Highest antigen sequence indentity to the following orthologs: Mouse - 64%, Rat - 57%.
This gene encodes an enzyme that converts the pancreatic proenzyme trypsinogen to trypsin, which activates other proenzymes including chymotrypsinogen and procarboxypeptidases. The precursor protein is cleaved into two chains that form a heterodimer linked by a disulfide bond. This protein is a member of the trypsin family of peptidases. Mutations in this gene cause enterokinase deficiency, a malabsorption disorder characterized by diarrhea and failure to thrive. [provided by RefSeq, Jul 2008].
Specifications
| TMPRSS15 | |
| Polyclonal | |
| Unconjugated | |
| TMPRSS15 | |
| enterokinase; enterokinase catalytic subunit; Enterokinase light chain; enteropeptidase; Enteropeptidase catalytic light chain; Enteropeptidase non-catalytic heavy chain; Entk; protease, serine, 7 (enterokinase); PRSS7; Serine protease 7; Tmprss15; Transmembrane protease serine 15; transmembrane protease, serine 15 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5651 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P98073 | |
| TMPRSS15 | |
| Recombinant protein corresponding to Human TMPRSS15. Recombinant protein control fragment (Product #RP-108260). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction