missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMPRSS11E Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548775
This item is not returnable.
View return policy
Description
TMPRSS11E Polyclonal Antibody for Western Blot
Serine protease which possesses both gelatinolytic and caseinolytic activities. Shows a preference for Arg in the P1 position.
Specifications
| TMPRSS11E | |
| Polyclonal | |
| Unconjugated | |
| TMPRSS11E | |
| DESC1; Serine protease DESC1; TMPRSS11E; TMPRSS11E2; transmembrane protease serine 11E; Transmembrane protease serine 11E catalytic chain; Transmembrane protease serine 11E non-catalytic chain; Transmembrane protease serine 11E2; transmembrane protease, serine 11E; transmembrane protease, serine 11E2; UNQ742/PRO1461 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 28983 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9UL52 | |
| TMPRSS11E | |
| KLH conjugated synthetic peptide between 249-277 amino acids from the Central region of human TMPRSS11E. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction