missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMPRSS11A Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-41140
This item is not returnable.
View return policy
Description
TMPRSS11A Polyclonal specifically detects TMPRSS11A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| TMPRSS11A | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| TMPRSS11A | |
| Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human ECRG1. The immunogen is located within amino acids 300 - 350 of ECRG1. Amino Acid Squence: YGGESQNDLREARVKI | |
| 0.1 mg | |
| Primary | |
| At least two isoforms of ECRG1 are known to exist; ECRG1 antibody is predicted to not cross-react with other ECRG family members. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1 ug/ml, ELISA | |
| Airway Trypsin-Like Protease 1, EC 3.4.21, EC 3.4.21., ECRG1, Epidermal Type II Transmembrane Serine Protease, Epidermal Type-II Transmembrane Serine Protease, Esophageal Cancer-Susceptibility Gene 1 Protein, HATL1, HESP, Transmembrane Protease Serine 11A, Transmembrane Protease, Serine 11A | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| 339967 | |
| Human, Mouse, Rat | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction