missing translation for 'onlineSavingsMsg'
Learn More

TMPA, MedChemExpress

Product Code. 16493537
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493537 50 mg 50mg
16467518 10 mg 10mg
16459997 5 mg 5mg
16420306 2 mg 2mg
16417476 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493537 Supplier Medchem Express Supplier No. HY18555
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress TMPA is an antagonist of nuclear receptor Nur77 and LKB1 interaction.

TMPA is an antagonist of nuclear receptor Nur77 and LKB1 interaction.

Specifications

Chemical Name or Material TMPA
CAS 1258275-73-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H32O6
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (262.83 mM)
SMILES COC1=C(CC(OCC)=O)C(C(CCCCCCC)=O)=CC(OC)=C1OC
Molecular Weight (g/mol) 380.48
Formula Weight 380.48
Percent Purity 98.56%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.