missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMP21 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12838502
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12838502 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12838502 Supplier Invitrogen™ Supplier No. PA520316

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is mouse brain tissue lysate. PA5-20316 can be used with blocking peptide PEP-0436.

TMP21 is a ubiquitously expressed protein that is involved in vesicular targeting and protein transport. More recent experiments have shown that it is also a component in the presenilin complex and modulates the -gamma-secretase but not the epsilon-secretase cleavage activity of the amyloid precursor protein. The presenilin complex is composed of the proteins APH1, nicastrin, and PEN2 in addition to presenilin-1. Together, these proteins cleave the amyloid precursor protein at what is known as the -gamma- and epsilon-sites and can lead to the accumulation of the Abeta cleavage product that is associated with Alzheimer's disease. Co-immunoprecipitation experiments using antibodies against these proteins also yielded TMP21 indicating that TMP21 may play a role in the regulation of this complex. Suppression of TMP21 expression by siRNA in transfected cells caused increased -gamma-secretase activity but not epsilon-secretase activity, and increased Abeta production, demonstrating that TMP21 can modulate -gamma-secretase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMP21
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TMED10
Gene Accession No. P49755, Q63584, Q9D1D4
Gene Alias 1110014C03Rik; 21 kDa transmembrane trafficking protein; 21 kDa transmembrane-trafficking protein; hypothetical protein; integral membrane protein p23; integral membrane protein Tmp21-I (p23); p23; p24 family protein delta-1; P24(DELTA); p24d1; p24delta; p24delta1; S31I125; S31III125; testicular tissue protein Li 206; tmed10; tmed10.L; TMP21; Tmp-21-I; transmembrane emp24 domain-containing protein 10; transmembrane emp24-like trafficking protein 10; transmembrane emp24-like trafficking protein 10 (yeast); transmembrane p24 trafficking protein 10; transmembrane p24 trafficking protein 10 L homeolog; transmembrane protein; transmembrane protein tmp21; transmembrane trafficking protein; transmembrane trafficking protein 21; XELAEV_18000191mg; Xp24d2; Xp24delta2
Gene Symbols TMED10
Host Species Rabbit
Immunogen An 18 amino acid peptide from near the center of human TMP21.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10972, 68581, 84599
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.