missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMOD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103052
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TMOD2.
This gene encodes a neuronal-specific member of the tropomodulin family of actin-regulatory proteins. The encoded protein caps the pointed end of actin filaments preventing both elongation and depolymerization. The capping activity of this protein is dependent on its association with tropomyosin. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
| TMOD2 | |
| Polyclonal | |
| Unconjugated | |
| Tmod2 | |
| neural tropomodulin; neuronal tropomodulin; NTMOD; N-TMOD; TMOD2; tropomodulin 2; tropomodulin 2 (neuronal); tropomodulin-2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 29767, 58814 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P70566, Q9NZR1 | |
| Tmod2 | |
| A synthesized peptide derived from human TMOD2(Accession Q9NZR1), corresponding to amino acid residues L36-F86. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction