missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMOD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578201
This item is not returnable.
View return policy
Description
Positive Control: U87-MG, SK-N-SH, IMR32, SK-N-AS Predicted Reactivity: Mouse (100%), Rat (100%), Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TMOD1 gene encodes a member of the tropomodulin family. The encoded protein is an actin-capping protein that regulates tropomyosin by binding to its N-terminus, inhibiting depolymerization and elongation of the pointed end of actin filaments and thereby influencing the structure of the erythrocyte membrane skeleton. Multiple transcript variants encoding the same protein have been found for this gene.
Specifications
| TMOD1 | |
| Polyclonal | |
| Unconjugated | |
| TMOD1 | |
| D9S57E; E tropomodulin 1; EGK_07519; Erythrocyte tropomodulin; ETMOD; E-Tmod; e-tropomodulin; Tmod; TMOD1; tropomodulin 1; tropomodulin-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7111 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1.27 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P28289 | |
| TMOD1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Tropomodulin 1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction