missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMLHE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16621-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes the protein trimethyllysine dioxygenase which is the first enzyme in the carnitine biosynthesis pathway. Carnitine play an essential role in the transport of activated fatty acids across the inner mitochondrial membrane. The encoded protein converts trimethyllysine into hydroxytrimethyllysine. A pseudogene of this gene is found on chromosome X. Alternate splicing results in multiple transcript variants.Specifications
| TMLHE | |
| Polyclonal | |
| Unconjugated | |
| TMLHE | |
| BBOX2, TML dioxygenase, TML hydroxylase, TMLD, TMLH, TMLHE, XAP130 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 170898, 192289, 55217 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q91ZE0, Q91ZW6, Q9NVH6 | |
| Tmlhe | |
| TMLHE Fusion Protein Ag9919 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction