missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMIGD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81844
This item is not returnable.
View return policy
Description
TMIGD1 Polyclonal specifically detects TMIGD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TMIGD1 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| TMIGD1 | |
| TMIGD1 antibody was raised against an 18 amino acid peptide near the center of human TMIGD1. The immunogen is located within amino acids 150 - 200 of TMIGD1. Amino Acid Squence: LDLEKSRHQIQQTSESFQ | |
| Affinity Purified | |
| RUO | |
| 388364 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin | |
| AWKS9372, TMIGD, transmembrane and immunoglobulin domain containing 1, transmembrane and immunoglobulin domain-containing protein 1, UNQ9372 | |
| Rabbit | |
| 24 kDa | |
| 0.1 mg | |
| Primary | |
| TMIGD1 antibody is human, mouse and rat reactive. At least two isoforms of TMIGD1 are known to exist; this antibody will detect both isoforms. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction