missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TMIGD1 Polyclonal specifically detects TMIGD1 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | TMIGD1 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | AWKS9372, TMIGD, transmembrane and immunoglobulin domain containing 1, transmembrane and immunoglobulin domain-containing protein 1, UNQ9372 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant TMIGD1 (NP_996663.1; Met1-Pro220). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?