missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMF1 Recombinant Rabbit Monoclonal Antibody (JE65-73), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA544831
This item is not returnable.
View return policy
Description
Adapter protein that may provide indirect link between the endocytic membrane traffic and the actin assembly machinery. May regulate the formation of clathrin-coated vesicles. Isoform 1 could be involved in brain-specific synaptic vesicle recycling.Specifications
TMF1 | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P82094 | |
TMF1 | |
Recombinant protein within human TMF aa 994-1,093/1,093. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
JE65-73 | |
Unconjugated | |
TMF1 | |
7030402D04Rik; Androgen receptor coactivator 160 kDa protein; androgen receptor-associated protein of 160 kDa; ARA160; Gm153; TATA element modulatory factor; TATA element modulatory factor 1; TMF; TMF1; Tmf1` | |
Rabbit | |
Protein A | |
RUO | |
7110 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |